missing translation for 'onlineSavingsMsg'
Learn More

Olive Oil, Pure, Fisher Chemical™

Brand:  Fisher Chemical O/0250/17

Code : 21

Additional Details : CAS Number : 8001-25-0 Weight : 2.50000kg

Quantity 2.5L

 View more versions of this product

Product Code. 10020270

  • 167.00 € / 2.50 L
Estimated Shipment: 01-06-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

8001-25-0
131764

Specifications

8001-25-0
0°C
Amber glass bottle
Viscous liquid Liquid
Yellow
131764
2.5L
Videos
SDS Documents
Certificates
Special Offers

Special Offers