missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole, 98%, Thermo Scientific™

A proton pump inhibitor

56.70 € - 141.00 €

Chemical Identifiers

CAS 73590-58-6
Molecular Formula C17H19N3O3S
Molecular Weight (g/mol) 345.417
MDL Number MFCD00083192
InChI Key SUBDBMMJDZJVOS-UHFFFAOYSA-N
Synonym omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
PubChem CID 4594
ChEBI CHEBI:77260
IUPAC Name 6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15465159
SDS View Documents
Thermo Scientific Alfa Aesar
J62860.03
1g
56.70 €
1g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15475159
SDS View Documents
Thermo Scientific Alfa Aesar
J62860.06
5g
141.00 €
5g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Omeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.

Solubility
Soluble in water (0.5 mg/ml), DMSO (25 mg/ml), and ethanol (4.5 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

73590-58-6
345.417
SUBDBMMJDZJVOS-UHFFFAOYSA-N
4594
6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
C17H19N3O3S
MFCD00083192
omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
CHEBI:77260
CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC

Specifications

73590-58-6
MFCD00083192
omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
SUBDBMMJDZJVOS-UHFFFAOYSA-N
6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
4594
345.42
Omeprazole
C17H19N3O3S
14,6845
Soluble in water (0.5mg/ml),DMSO (25mg/ml),and ethanol (4.5mg/ml).
CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC
345.417
CHEBI:77260
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'rtecsNumber' : DD9087000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.