Learn More
Julabo™ Open heating bath circulator, CORIO™ CD-B5
Brand:  Julabo™ 9012405
Additional Details : Weight : 8.20000kg
Product Code. 15450572
  • 1755.00 € / Each
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
The Open heating bath circulators of the new CORIO™ series feature high-quality bath tanks made of stainless steel and pump connections.
  • Model for internal and external applications
  • Bright, white, easy-to-read display
  • Very quiet
  • Easy pump change-over between internal and external circulation
  • USB connection
  • High-quality bath tanks made of stainless steel
Ideal for temperature control of samples, preparation of samples for serology, and clinical chemistry, analysis, material testing, and more. External temperature control applications in combination with measuring cells, photometers, refractometers, polarimeters, and more.
Working temperature range: 20°C to 150°CTemperature stability: ±0.03°CHeating capacity: 2kWPump capacity flow rate: 15L/minDimensions W x L x H (cm): 23 x 38 x 41Filling volume (L): 3.5 to 5