missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jaytec Glass™ Open Top Borosilicate Glass Dropping Funnels
Open Top Borosilicate Glass Dropping Funnels for easy refilling in situ. The stopcock permits carefully controlled flow of reagents.
33.35 € - 41.25 €
Specifications
Product Type | Dropping funnel |
---|---|
Material | Borosilicate Glass |
Graduated | No |
Includes | PTFE stopcock key |
Stopcock | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12338577
|
Jaytec Glass™
FTP 050 |
50 mL |
34.00 €
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
12328577
|
Jaytec Glass™
FPM-210-030D |
25 mL |
33.35 €
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
12348577
|
Jaytec Glass™
FPM-210-070X |
100 mL |
37.05 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
12358577
|
Jaytec Glass™
FTP250 |
250 mL |
41.25 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
- Open top
- Cylindrical shape
- With interchangeable PTFE stopcock
Specifications
Dropping funnel | |
No | |
Yes |
Borosilicate Glass | |
PTFE stopcock key |