Learn More
Thermo Scientific™ Open Top Screw Caps for Sample Storage Vials
Open Top Screw Caps for Sample Storage Vials
7.95 € - 12.48 €
Specifications
Closure Material | Polypropylene |
---|---|
Closure Type | Screw Cap (Open-Top) |
Quantity | 100 |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
12468626
|
Thermo Scientific™
B7807-20 |
Black; 20-400 |
11.78 €
Pack of 100 |
In Stock
Log in to see stock. |
|||||
12478626
|
Thermo Scientific™
B7807-24 |
White; 24-400 |
12.48 €
Pack of 100 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
13110993
|
Thermo Scientific™
B7807-15 |
Black; 15-425 |
7.95 €
Pack of 100 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
12623735
|
Thermo Scientific™
B7807-8 |
Black; 8-425 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13171663
|
Thermo Scientific™
B7807-13 |
Black; 13-425 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12458626
|
Thermo Scientific™
B7807-18 |
Black; 18-400 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Open Top Screw Caps for Sample Storage Vials
Specifications
Polypropylene | |
Screw Cap (Open-Top) | |
100 |