Learn More
BRAND™ Optical Glass Cavity Slides
With rectangular ground edges.
Brand:  BRAND™ 475505
Code : NEW
Additional Details : Weight : 0.28576kg
 View more versions of this product
Product Code. 10555602
  • 180.00 € / Pack of 50
Estimated Shipment: 15-04-2024
to see stock.
Add to basket
Description
Description
  • White (clear) soda-lime glass, hydrolytic class 3
  • Single concavity
  • Provided in packs of 50
Specifications
Specifications
Ground
Soda Lime Glass
1.2 to 1.5 mm
Clear White
Optical Glass Slide
1
50