missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ 5 Performance GC Capillary Column

Brand:  Macherey-Nagel™ 726809.60

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10052991

Quantity Price
1 1091.40 € / Each
3 916.24 € / Each
Estimated Shipment: 04-01-2022

Add to basket


Description

Description

Specifications

Specifications

0.5 mm
0.32 mm
Optima™ 5
340°C
60 m
1 Each
High Performance GC Capillary Column
GC
G27
3 µm
Optima™ 5
Videos
SDS
Documents
Certificates
Special Offers

Special Offers