missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ HinfI, Fisher BioReagents™
Brand: Fisher Bioreagents BP8051-1
Code : 32
Additional Details : Weight : 0.01000kg
Description
5'...G^A N T C...3'
3'...C T N A^G...5'
Supplied with: 10X OPTIZYME Buffer 5
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 5: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 0 - 20%
- Buffer 2: 20 - 50%
- Buffer 3: 50 - 100%
- Buffer 4: 50 - 100%
- Buffer 5: 100%
- 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
- 5'...GANTm5C G...3'
- 5'...m5C GANTm5C G...3'
- 5'...TGm6ANTC(N)5 TGCT...3'
Specifications
10 U/μL | |
37°C | |
10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol | |
2000 U | |
HinfI |
25 to 50% Water, >50% Glycerin | |
>95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with HaeIII | |
Keep container tightly closed | |
G.ANTC | |
Liquid |