Learn More
Thermo Scientific™ Orion™ 8102BN ROSS™ Combination pH Electrode
Get stable readings to 0.01 pH in less than 30 seconds with this combination pH electrode.
279.00 € - 374.00 €
Specifications
Connector Types | WPBNC |
---|---|
Junction | Ceramic |
Material | Glass Body |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 100°C |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10615261
|
Thermo Scientific™
8102BN |
BNC connector |
279.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
10282921
|
Thermo Scientific™
8102SC |
Screw cap connector |
374.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Get stable readings to 0.01 pH in less than 30 seconds with the Thermo Scientific™ Orion™ 8102BN ROSS™ Combination pH Electrode. This electrode provides results three to five times more precise than those obtained with conventional electrodes.
Features:
- Works in extreme case of samples varying from one another by 50°C or more
- Drift is less than 0.002 pH per day, restandardization is minimized
- General purpose, top performance combination electrode
- Double junction
- TRIS compatible
- Waterproof BNC connector
Includes:
- One bottle 810007 fill solution
Requires:
- pH buffers for calibration
- Beakers, plastic or glass
- Magnetic stirrer or Orion stirrer probe
- Distilled or deionized water
Recommended for:
- Quality Control (QC) and research applications
Warranty: 12 months from date of purchase by Buyer or 18 months from date of shipment from Seller, whichever is earlier
Specifications
WPBNC | |
Glass Body | |
0°C to 100°C | |
3M KCl | |
ROSS | |
Short-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode. | |
4.72 in. | |
120 mm | |
0.47 in. | |
16 mm |
Ceramic | |
0 to 14 | |
<0.002pH/day | |
One 60mL bottle of Orion ROSS filling solution and spout cap | |
0.01 | |
32°F to 212°F | |
39.37 in. | |
1 m | |
12 mm |