missing translation for 'onlineSavingsMsg'
Learn More

ORP buffer +475mV, Hamilton™

Catalog Number 11543004
Click to view available options
Quantity:
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Color Yellow
pH 1.1
Physical Form Liquid
Quantity 250 mL
Solution Type Buffer
Formulation 2% Diammonium iron bis(sulphate)-6-hydrate, 0.5% Hydrochloric acid
Odor Odorless
Solubility Information Water Soluble
compliance-icons
Product Identifier
  • redox buffer 475
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : diammonium iron bis(sulphate)-6-hydrate,hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'