Learn More
Orthophosphoric Acid, 85%, HPLC for Electrochemical Detection, Certified, Fisher Chemical™
H3O4P, CAS Number-7664-38-2, acide phosphorique, evits, acido fosforico, white, acidum phosphoricum, sonac, o-phosphoric acid, wc-reiniger, orthophosphoric acid, phosphorsaeure, 500mL, 158 deg.C, CHEBI:26078, Undesignated, NBIIXXVUZAFLBC-UHFFFAOYSA-N, 21 deg.C, 11340, 97.994
Brand: Fisher Chemical O/0515/PB08
67.24 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 25
Additional Details : CAS Number : 7664-38-2 Weight : 0.84000kg Transport : UN number : 1805 Chem class : 8 Pack group : III
Description
H3O4P, CAS Number-7664-38-2, acide phosphorique, evits, acido fosforico, white, acidum phosphoricum, sonac, o-phosphoric acid, wc-reiniger, orthophosphoric acid, phosphorsaeure, 500mL, 158 deg.C, CHEBI:26078, Undesignated, NBIIXXVUZAFLBC-UHFFFAOYSA-N, 21 deg.C, 11340, 97.994
Specifications
158°C | |
7664-38-2 | |
Liquid | |
H3O4P | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 | |
2 hPa at 20°C |
21°C | |
2 | |
500 mL | |
11340 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
Plastic Coated Glass bottle | |
Vapor Density: 3.4 |
Safety and Handling
- Phosphoric acid, 85+% solution in water
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Orthophosphoric acid