missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Oval Stirring Bars
Provide effective stirring of samples via magnetic manipulation
Brand: Kartell™ 0068400
Additional Details : Weight : 0.06500kg
Description
- Ideal for containers or bottles with round bottom
- Suitable for foodstuff
- magnet with PTFE coating
- Oval shape is ideal for ensuring homogeneous mixing in round-bottomed beakers
Specifications
12 mm | |
Magnet, PTFE | |
25 mm |
10/Pk. | |
Ideal for containers/ bottles with round bottoms | |
Oval |