Learn More
MP Biomedicals™ Ovalbumin, 5g
Brand: MP Biomedicals™ 08950512
Code : NEW
Additional Details : CAS Number : 9048-46-8 Weight : 0.01890kg
Specifications
Immunoassay | |
5g | |
-20°C | |
Lyophilized |
Ovalbumin from Chicken Egg | |
Chicken Egg | |
Chicken |