missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ Blotting Paper
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with this high-quality blotting paper.
36.20 € - 277.00 €
Specifications
Packaging Type | Sheets |
---|---|
Flow Rate | 35 mL/min. |
For Use With (Equipment) | Owl electroblotting systems |
Weight | 183 g/m² |
Format | Sheet |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
12484627
|
Thermo Scientific™
FP-1 |
20 cm | 20 cm |
82.70 €
Pack of 100 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
12494627
|
Thermo Scientific™
FP-2 |
35 cm | 45 cm |
161.00 €
Pack of 100 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
12404637
|
Thermo Scientific™
FP-3 |
46 cm | 57 cm |
277.00 €
Pack of 100 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
12414637
|
Thermo Scientific™
FP-4 |
10 cm | 10 cm |
36.20 €
Pack of 100 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
||||
12424637
|
Thermo Scientific™
FP-5 |
18 cm | 20 cm |
82.35 €
Pack of 100 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
11932985
|
Thermo Scientific™
FP-6 |
9 cm | 9 cm |
49.05 €
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
12444637
|
Thermo Scientific™
FP-7 |
12 cm | 16 cm |
74.15 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.
- High purity, 100% cotton fiber
- Available in pre-cut mini gel and large format gel sizes
- Compatible with Owl electroblotting systems
Ordering Information:
- Sold in packs of 100 sheets
Specifications
Sheets | |
Owl electroblotting systems | |
Sheet | |
Owl |
35 mL/min. | |
183 g/m² | |
100% Cotton Fiber |