missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxaliplatin, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Oxaliplatin
Specifications
Specifications
| Chemical Name or Material | Oxaliplatin |
| CAS | 61825-94-3 |
| Analyte or Component Names | Oxaliplatin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H14 N2 O4 Pt |
| Certifications/Compliance | ISO 17034 |
| Synonym | Oxaliplatin, (SP-4-2)-[(1R,2R)-Cyclohexane-1,2-diamine-kappaN,kappaN′][ethanedioato(2-)-kappaO1,kappaO2]platinum |
| InChI Formula | InChI=1S/C6H14N2.C2H2O4.Pt/c7-5-3-1-2-4-6(5)8;3-1(4)2(5)6;/h5-6H,1-4,7-8H2;(H,3,4)(H,5,6);/q;;+2/p-2/t5-,6-;;/m1../s1 |
| SMILES | O=C1C(=O)[O-][Pt+2]2([NH2][C@@H]3CCCC[C@H]3[NH2]2)[O-]1 |
| IUPAC Name | (SP-4-2)-[(1R,2R)-Cyclohexane-1,2-diamine-κN,κN′][ethanedioato(2-)-κO1,κO2]platinum |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction