missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxiracetam, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Oxiracetam
Specifications
Specifications
| Chemical Name or Material | Oxiracetam |
| CAS | 62613-82-5 |
| Analyte or Component Names | Oxiracetam |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H10 N2 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2-(4-Hydroxy-2-oxopyrrolidin-1-yl)acetamide, 4-Hydroxy-2-oxo-1-pyrrolidineacetamide, 4-Hydroxypyrrolidone-2-acetamide, CGP 21690, CGP 21690E, CT 848, Hydroxypiracetam, ISF 2522, Neuractiv, Neuromet, Oxiracetam |
| InChI Formula | InChI=1S/C6H10N2O3/c7-5(10)3-8-2-4(9)1-6(8)11/h4,9H,1-3H2,(H2,7,10) |
| SMILES | NC(=O)CN1CC(O)CC1=O |
| IUPAC Name | 2-(4-hydroxy-2-oxopyrrolidin-1-yl)acetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction