Learn More
Thermo Scientific™ Slanetz and Bartley Medium (Dehydrated)
Detect and enumerate enterococci from water and food samples with this selective medium.
Brand: Thermo Scientific™ CM0377B
170.05 EUR valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Code : 64
Additional Details : Weight : 0.59200kg
Description
Detect and enumerate enterococci from water and food samples with Thermo Scientific™ Slanetz and Bartley Medium (Dehydrated). Slanetz and Bartley1 originally devised this medium to detect and enumerate enterococci by the technique of membrane filtration, but it has also proved useful as a direct plating medium2, 3. The medium is very selective for enterococci and, when it is incubated at elevated temperatures (44-45°C), all red or maroon colonies may be accepted as presumptive enterococci4, 5.
The detection and enumeration of enterococci is of primary importance for food hygiene monitoring and microbial quality of water and food.
The Environment Agency ‘Microbiology of Drinking Water 2002’6 recommend the use of Slanetz and Bartley medium for the enumeration of enterococci in water supplies, as does ISO in the standard for water quality7. Food samples can be examined for enterococci by the method suggested by the Nordic Committee of Food Analysis3.
- Easy-to-read: due to the reduction of tetrazolium chloride in the medium the enterococci form dark red/ maroon colonies.
- Selective: sodium azide in the medium is selective against Gram-negative organisms.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Industrial Reference: NMKL. | |
Powder | |
500 g |
Slanetz and Bartley Medium (Enterococcus Agar, Dehydrated) | |
Dehydrated Culture Media | |
For 11.9L medium |
Safety and Handling
- SLANETZ and BARTLEY MEDIUM
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Sodium azide
For Laboratory Use Only