missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tergitol-7 Agar (Dehydrated)

Enumerate and differentiate coliforms from water and food samples with this selective, differential medium when used with TTC Solution Supplement.

Brand:  Thermo Scientific™ CM0793B

Code : 64

Additional Details : Weight : 1.90510kg

Product Code. 11973732

  • 145.00 € / 500g
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

Enumerate and differentiate coliforms from water and food samples with selective Thermo Scientific™ Oxoid™ Tergitol-7 Agar (Dehydrated) when used with TTC Solution Supplement (SR0148). Tergitol 7 inhibits Gram-positive organisms allowing superior recovery of coliforms. Fermentation of lactose is seen by a change in color of the pH indicator bromothymol blue. Tri-phenyltetrazolium chloride (TTC) is rapidly reduced to red formazan by most coliform organisms except Escherichia coli and Enterobacter aerogenes allowing easy differentiation.

Tergitol-7 Agar is based on the formulation described by Chapman1 and is recommended for the selective isolation and differentiation of the coliform group.

  • Easy-to-read: easily differentiate Escherichia coli (yellow colonies) and Enterobacter species (greenish/yellow colonies) from coliforms (red colonies).
  • Selective: Tergitol-7 inhibits Gram positive organisms and minimizes the swarming of Proteus, allowing superior recovery of coliforms.
  • Recommended: for examination of foodstuffs for fecal contamination2.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Specifications

Industrial Reference: NMKL, ISO.
Powder
500 g
Tergitol-7 Agar Base
Agar
For 9.2L medium
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

For Laboratory Use Only