Learn More
Thermo Scientific™ Oxoid™ TTC Solution
Oxoid TTC Solution (0.125%) is used for the enumeration and identification of coliform bacteria.
Brand: Thermo Scientific™ SR0148A
52.99 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : 65
Additional Details : Weight : 0.16200kg
Description
Oxoid TTC Solution (0.125%) is used for the enumeration and identification of coliform bacteria.
Add to Tergitol 7 Agar, Part No. CM0793BSpecifications
TTC Solution | |
Solution | |
Each vial for 100mL medium |
Liquid | |
10 x 2 mL |