missing translation for 'onlineSavingsMsg'
Learn More

Oxytetracycline hydrochloride, Thermo Scientific™

Impairs mitochondrial protein synthesis

83.60 € - 263.00 €

Chemical Identifiers

CAS 2058-46-0
Molecular Formula C22H25ClN2O9
Molecular Weight (g/mol) 496.897
MDL Number MFCD00135815
InChI Key UTAHTGPLLFLKGW-IFLJXUKPSA-N
PubChem CID 121235537
IUPAC Name (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15464869
SDS View Documents
Thermo Scientific Alfa Aesar
J62489.14
25g
83.60 €
25g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15474869
SDS View Documents
Thermo Scientific Alfa Aesar
J62489.22
100g
263.00 €
100g
View alternative products
Save Up to 
»
Estimated Shipment: 25-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Oxytetracycline is an antibiotic indicated for treatment of infections caused by Gram positive and Gram negative microorganisms such as Mycoplasma pneumoniae, Pasteurella pestis, Escherichia coli, Haemophilus influenzae, and Diplococcus pneumoniae. It is used in studies on the oxytetracycline-resistance gene (otrA). Oxytetracycline hydrochloride is used to study phagosome- lysosome (P-L) fusion in P388D1 cells1 and antibiotic susceptibilities of Mycoplasma bovis isolates.

Solubility
Soluble in ethanol

Notes
Concentrated aqueous solutions at neutral pH will hydrolyze oxytetracycline and cause it to precipitate
Specifications

Chemical Identifiers

2058-46-0
496.897
UTAHTGPLLFLKGW-IFLJXUKPSA-N
(4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
C22H25ClN2O9
MFCD00135815
121235537
CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl

Specifications

2058-46-0
Air and light sensitive
MFCD00135815
14,6976
UTAHTGPLLFLKGW-IFLJXUKPSA-N
(4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
121235537
Oxytetracycline hydrochloride
Gray to Yellow
C22H25ClN2O9
3853107
Soluble in ethanol
CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl
496.897
496.9
Videos
Safety and Handling

Safety and Handling

P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c

H319-H361d

missing translation for 'einecsNumber' : 218-161-2

missing translation for 'rtecsNumber' : QI8225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Product Identifier
  • Oxytetracycline hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: Oxytetracycline hydrochloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.