missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluidine, 99%, crystalline molten mass, Thermo Scientific Chemicals
17.13 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Glass Bottle
Metal drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 41.0°C to 46.0°C |
Color | White to Brown |
Boiling Point | 200.0°C |
Flash Point | 87°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | CH3C6H4NH2 |
Quantity | 250 g |
Merck Index | 15,9696 |
Show More |
Safety and Handling
Product Identifier
- p-Toluidine
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H311-Toxic in contact with skin.
- H351-Suspected of causing cancer.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'