missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Tumor Suppressor Protein Rabbit, Clone: TP53/1799R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 7157-RBM10-P1
Additional Details : Weight : 0.01000kg
Specifications
p53 Tumor Suppressor Protein | |
Predominantly nuclear | |
TP53/1799R | |
Unconjugated | |
Rabbit | |
MDA-MB-231 Cells. Breast or Colon carcinoma | |
100 μg | |
Primary | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
Recombinant full-length human TP53 protein | |
Protein A/G Purified | |
RUO | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |