Learn More
Paclitaxel, 99.5+%, Thermo Scientific™
Paclitaxel (CAS # 33069-62-4) is a microtubule inhibitor produced from the extract of the Pacific yew tree.
153.00 € - 723.00 €
Chemical Identifiers
| CAS | 33069-62-4 |
|---|---|
| Molecular Formula | C47H51NO14 |
| Molecular Weight (g/mol) | 853.92 |
| MDL Number | MFCD00869953 |
| InChI Key | RCINICONZNJXQF-VAZQATRQSA-N |
| Synonym | paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel |
| PubChem CID | 133640187 |
| IUPAC Name | (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate |
| SMILES | CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15495039
|
Thermo Scientific Alfa Aesar
J62734.MC |
100mg |
153.00 €
100mg |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15405049
|
Thermo Scientific Alfa Aesar
J62734.ME |
500mg |
446.00 €
500mg |
Estimated Shipment: 21-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15485039
|
Thermo Scientific Alfa Aesar
J62734.03 |
1g |
723.00 €
1g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
- Paclitaxel is a tetracyclic diterpenoid isolated from the Pacific yew, Taxus brevifolia
- Paclitaxel acts as an antineoplastic agent by binding tubulin and preventing cell division
- Paclitaxel may also bind Bcl-2 and lead to disinhibition of apoptosis
Application
- Paclitaxel is used in the study of lung, breast, and ovarian cancers; it may be useful in the research of other cancer types
- It is suitable for cell culture, biochemistry, and molecular biology applications
Chemical Identifiers
| 33069-62-4 | |
| 853.92 | |
| RCINICONZNJXQF-VAZQATRQSA-N | |
| 133640187 | |
| CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
| C47H51NO14 | |
| MFCD00869953 | |
| paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel | |
| (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate |
Specifications
| 33069-62-4 | |
| MFCD00869953 | |
| 14,6982 | |
| Soluble in DMSO (200mg/ml) or ethanol (40mg/ml); Soluble in dimethylsulfoxide,methanol,ethanol,acetonitrile,cremophor and dimehtylsulfamide; Sparingly soluble in aqueous buffer. | |
| CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 853.92 | |
| 853.91 | |
| Paclitaxel |
| C47H51NO14 | |
| 1420457 | |
| paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel | |
| RCINICONZNJXQF-VAZQATRQSA-N | |
| (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate | |
| 133640187 | |
| ≥99.5% |
Safety and Handling
GHS H Statement
H334-H341-H361-H318-H302-H312-H332-H335-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P342+P311-P363-P501c
H302+H312+H332-H318-H334-H335-H361
missing translation for 'rtecsNumber' : DA8340700
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
- Paclitaxel
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H361-Suspected of damaging fertility or the unborn child.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contain: Paclitaxel
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.