missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on activated carbon powder, Type A405023-5, nominally 50% water wet

Brand:  Alfa Aesar™ 045051.14

Code : NEW

Additional Details : Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15473935

  • 224.00 € / 25g

Estimated Shipment date
from Supplier 17-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sold on a dry weight basis
Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]

Specifications

Palladium
7440-05-3
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Powder
25g
5% on activated carbon powder, Type A405023-5, Nominally 50% water wet
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
Odorless
Videos
SDS Documents