missing translation for 'onlineSavingsMsg'
Learn More

Palladium nitrate & Magnesium nitrate, Matrix Modifier Solution, Specpure™

Product Code. 15453965
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 15453965

Brand: Thermo Scientific Alfa Aesar 045290.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Concentration 0.075% Pd & 0.05% Mg(NO3)2 in 2% HNO3
MDL Number MFCD09910543
UN Number UN3264
Quantity 250 mL
Grade Specpure™
Chemical Name or Material Palladium nitrate & Magnesium nitrate Matrix Modifier Solution

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.