missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Particle Certified Glass Containers
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2500 mL
4000 mL
Description:
Amber glass Boston round bottle
Amber glass Boston round bottle with closed top closure
Amber glass Boston round bottle with open-top septa closure
Amber glass jug with closure
Amber glass wide mouth packer bottle with closure
Clear Glass Jar, Wide Mouth, with Polypropylene Closure
Clear Glass Jar, Wide mouth with Polypropylene Closure
Clear glass Boston round bottle with closed top closure
No. per Case:
4
12
24
Description
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Available in clear or amber
- Assembled with low-shedding polypropylene caps with chemically inert PTFE faced liners that do not contain adhesives
- Individual bottles double bagged to maintain integrity until point of use
- 4L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
Low particle applications
Specifications
Capacity | 4 oz., 125 mL |
Closure Liner | PTFE/Silicone |
Closure Material | Polypropylene, Low-shedding |
Closure Size | 24-414 mm |
Diameter (Metric) | 47.63 mm |
Height (English) | 4.375 in. |
Height (Metric) | 111.13 mm |
Capacity (Metric) | 125 mL |
Capacity (English) | 4 oz. |
Color | Amber |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'