missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX7 Mouse, Clone: PAX7/497, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 505.00 €
Specifications
Antigen | PAX7 |
---|---|
Clone | PAX7/497 |
Concentration | 200μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960608
|
EnquireBio
5081-MSM1-P1 |
100 μg |
505.00 €
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950608
|
EnquireBio
5081-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
PAX7 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
5081 | |
Recombinant fragment (aa301-505) of human PAX7 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
57kDa |
PAX7/497 | |
Western Blot | |
Unconjugated | |
RUO | |
NP_039236 | |
PAX7 | |
IgG1 κ | |
Protein A/G Purified | |
Nuclear | |
HeLa, Jurkat, A431, MCF-7, U2OS or NIH/3T3 cells. Rhabdomyosarcoma (RMS) |