missing translation for 'onlineSavingsMsg'
Learn More

PAX8 Mouse, Clone: PAX8/1491, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 7849-MSM1-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961108

  • 485.00 € / 100µg
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PAX8
Nuclear
PAX8/1491
Unconjugated
XP_016860367
Mouse
62kDa
Protein A/G Purified
RUO
7849
IgG2b
Western Blot
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
PAX8
Recombinant human PAX8 fragment (aa60-261)
Raji cells. Renal Cell Carcinoma (RCC)
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers