missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PCR 12-Strip Caps
Manufactured from polypropyle without lubricants or chemical additives that can contaminate reagents and samples.PCR 12-Strip Caps are autoclavable and RNase-, DNase-, and DNA-free. Caps are domed and available in a variety of colors for sample identification.
55.80 € - 64.10 €
Specifications
For Use With (Equipment) | PCR tubes |
---|---|
Shape | Domed |
Material | Polypropylene |
Dnase Free | Yes |
RNase Free | Yes |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
10080902
|
BRAND™
781290 |
Clear |
55.80 €
Pack of 125 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
10090902
|
BRAND™
781292 |
Yellow |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10234902
|
BRAND™
781293 |
Green |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10001032
|
BRAND™
781294 |
Blue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.- Autoclavable
- RNase-, DNase-, and DNA-free
- All 12-Strip caps are domed and available in a variety of colors for sample identification
Specifications
PCR tubes | |
Polypropylene | |
Yes | |
Tube Strip Cap |
Domed | |
Yes | |
Autoclavable |