missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes

Product Code. 10398031 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.5 mL
1.5 mL
2 mL
Unit Size:
Pack of 1000
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10398031 1.5 mL Pack of 1000
10318661 0.5 mL Pack of 500
10073321 2 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10398031 Supplier Eppendorf™ Supplier No. 0030123328
  • ONLINE EXCLUSIVE PRICE
    61.85 €
    57.50 € / Pack of 1000
    Save 4.35 €
    7% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.

Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.

  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Guaranteed functionality from -86 to 100°C
  • Large frosted lid and surface on the side for easy labeling
  • Autoclavable when open (121°C, 20 min)
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Volume (Metric) 1.5 mL
Quantity 1000/Pk.
Material Polypropylene
Autoclavable Autoclavable
Closure Included Yes
Product Type Centrifuge Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.