Learn More
Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes

Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.
59.49 € - 83.70 €
Specifications
Color | Natural |
---|---|
Material | Polypropylene |
Autoclavable | Autoclavable |
Closure Included | Yes |
Product Type | Centrifuge Tube |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10318661
![]() |
Eppendorf™
0030123301 |
0.5 mL |
59.49 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10398031
![]() |
Eppendorf™
0030123328 |
1.5 mL |
61.27 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10073321
![]() |
Eppendorf™
0030123344 |
2 mL |
83.70 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.
- Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
- g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
- Precise lid sealing for minimal evaporation rates during long-term storage
- Guaranteed functionality from -86 to 100°C
- Large frosted lid and surface on the side for easy labeling
- Autoclavable when open (121°C, 20 min)
Specifications
Natural | |
Autoclavable | |
Centrifuge Tube |
Polypropylene | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.