missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PDHB 293T Cell Transient Overexpression Lysate (Denatured)

Human overexpression lysate

Brand:  Abnova™ H00005162-T01.100uL

Additional Details : Weight : 0.00010kg


Product Code. 16162565

  • 209.00 € / 100µL
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Western Blot
PDHB 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 39.2kDa
Transient overexpression cell lysate was tested with anti-PDHB antibody (Mfr. No. H00005162-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
293T
BC001924.1
pCMV-PDHB full-length
100 μL
Human
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100UL PDHB 293T Cell Transient Overexpression Lysa

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers