missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical PEEK Tubing, 360μm O.D., 5 ft
Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs
33.50 €
Specifications
pH Range | 2 to 14 |
---|---|
Diameter (Metric) Outer | 360 μm |
Overall Length (English) | 5 ft. |
Material | PEEK |
Max. Operating Temperature (Metric) | 100°C |
Product Code | Brand | Color | Diameter (English) Inner | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (English) Inner | Price | Quantity & Availability | ||||
15781049
|
Trajan Scientific and Medical
1304002000-5F |
Natural | 50 μm (0.002 in.) |
33.50 €
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15721059
|
Trajan Scientific and Medical
1304003006-5F |
Black | 75 μm (0.003 in.) |
33.50 €
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15761059
|
Trajan Scientific and Medical
1304004001-5F |
Red | 100 μm (0.004 in.) |
33.50 €
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15701069
|
Trajan Scientific and Medical
1304005005-5F |
Green | 125 μm (0.005 in.) |
33.50 €
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15741069
|
Trajan Scientific and Medical
1304006002-5F |
Yellow | 150 μm (0.006 in.) |
33.50 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
Description
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifications
2 to 14 | |
5 ft. | |
100°C |
360 μm | |
PEEK |
For research use only