missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ Optime™ III Ear Muffs

Catalog Number p-8006604
21.40 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.


21.40 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Color:
Black, Red
Green, Black
Style:
Headband
Helmet-mounted
Neckband

Developed for use in extremely high noise environments, particularly those dominated by high frequency sounds. 3M™ PELTOR™ Optime™ III Ear Muffs are based on technology featuring double cup design to help minimize resonance effect, resulting in excellent high frequency attenuation.

  • Modern, stylish cup design
  • Good speech intelligibility
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy-to-replace cushions and inserts help keep them hygienically clean
  • Unique low-profile headband design helps maintain constant pressure
  • Soft, wide cusions help reduce pressure around the ears and improve comfort and wearability
  • One size fits most
Color Black, Red
Certifications/Compliance CE Approved
Size One size fits most
For Use With (Application) Used in Extremely Noise Environments Like Airports, Marine Engine Rooms, Power Stations and Print Works
Plug Shape Oval
Product Type Ear Muffs Optime III
Style Headband
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'