missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ X5 Series Ear Muffs

Catalog Number p-6838003
35.10 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.


35.10 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Model:
X5A
Dielectric:
Yes
Style:
Over-the-head
This item is not returnable. View return policy
This item is not returnable. View return policy

PELTOR™ X5 Series Ear Muffs feature a combination of specially-formulated foam plus an innovative spacer and cup design to make it ideal for very high level noise situations.

  • Unparalleled attenuation without the need for double protection
  • Offered in over-the-head or hard hat-attached styles
  • Twin headband design helps reduce heat buildup with good fit and balance
  • Combination of new technologies allows for more room inside the ear cup for a more comfortable fit
  • Ear cup pivot points tilt for optimum comfort and efficiency
  • Dielectric (models X5A and X5P5E only)
  • Compatibility with hard hats (models X5P3E and X5P5E)

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Color Black
ANSI S3.1974 Yes
Cushion Material Foam
CSA Z94.2 Class AL
Cushion Color Black
Headband Material Wire
For Use With (Application) Composite Finishing, Grinding, Machine Operations, Paper Mills, Quarrying, Transportation, Welding
Model X5A
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 31 dB
Product Type X5 Series
Headband Color Black
Dielectric Yes
Material ABS/TPU, PU Foam and PVC
Style Over-the-head
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'