missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoyl chloride, 98%, ACROS Organics™

Brand:  Acros Organics 147121000

Code : 29

Additional Details : CAS Number : 2251-50-5 Weight : 0.15000kg Transport : UN number : 3265 Chem class : 8 Pack group : II


Quantity 100g

 View more versions of this product

Product Code. 11964981

  • 306.00 € / 100g

Estimated Shipment
26-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acylation reagent
Specifications

Chemical Identifiers

2251-50-5
230.52
pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate
CHEBI:39425
C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)Cl
C7ClF5O
MYHOHFDYWMPGJY-UHFFFAOYSA-N
75256
2,3,4,5,6-pentafluorobenzoyl chloride

Specifications

Pentafluorobenzoyl chloride
Authentic
1.4520 to 1.4540
Colorless to Yellow
2251-50-5
C7ClF5O
pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate
MYHOHFDYWMPGJY-UHFFFAOYSA-N
2,3,4,5,6-pentafluorobenzoyl chloride
75256
230.52
98%
1.6000g/mL
Glass Bottle
1.6
100g
97.5% min. (GC)
C6F5COCl
Solubility in water: decomposes.
C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)Cl
230.52
CHEBI:39425
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pentafluorobenzoyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents