missing translation for 'onlineSavingsMsg'
Learn More

Pentylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog Number 11935961
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
693-25-4, 60-29-7
C5H11BrMg
175.35
MFCD00000045
ZXWGTPYWPZGZPT-UHFFFAOYSA-M
amylmagnesium bromide, magnesium, bromopentyl, bromopentylmagnesium, n-pentylmagnesium bromide, pentylmagnesium bromide, pentylmagnesium bromide solution, n-pentyl magnesium bromide, npentmgbr;, pentylmagnesiumbromide, amyl-magnesium bromide, grignard reagent
101838
magnesium;pentane;bromide
CCCCC[Mg]Br
This item is not returnable. View return policy
693-25-4, 60-29-7
C5H11BrMg
175.35
MFCD00000045
ZXWGTPYWPZGZPT-UHFFFAOYSA-M
amylmagnesium bromide, magnesium, bromopentyl, bromopentylmagnesium, n-pentylmagnesium bromide, pentylmagnesium bromide, pentylmagnesium bromide solution, n-pentyl magnesium bromide, npentmgbr;, pentylmagnesiumbromide, amyl-magnesium bromide, grignard reagent
101838
magnesium;pentane;bromide
CCCCC[Mg]Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black to Yellow
Density 0.9470g/mL
Flash Point -40°C
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Specific Gravity 0.947
Solubility Information Solubility in water: reacts
Formula Weight 175.35
Physical Form Solution
Chemical Name or Material Pentylmagnesium bromide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pentylmagnesium bromide, 2M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'