Learn More
Thermo Scientific™ Oxoid™ Peptone Water (Bottled)
Grow wide variety of microorganisms with this non selective enrichment medium in convenient ready prepared bottles.
Brand: Thermo Scientific™ Oxoid™ BO0208B
Code : 64
Additional Details : Weight : 1.00000kg
Description
Grow a variety of microorganisms with Thermo Scientific™ Oxoid™ Peptone Water ready-prepared bottled medium. This non-selective broth medium is used as a primary enrichment medium for the growth of bacteria1. It can also be used as an inoculum for a series of fermentation tubes or other diagnostic media.
Use Peptone Water for growth of a wide variety of microorganisms.
- Ready-to-use: convenience of prepared media.
- Simple: easy-to-use with minimum training.
- Long shelf life: up to 12 months.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Straw, clear | |
Peptone Water (Bottled) | |
United Kingdom | |
3.0g + 0.3g | |
24/Pk. |
2 to 25°C | |
Bijou bottles | |
Peptone | |
Bijou bottles with plastic screw cap closures in a box |