missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctanesulfonamide, 95%, Thermo Scientific Chemicals

Catalog Number p-7077811
Click to view available options
Quantity:
1 g
5 g
754-91-6
C8H2F17NO2S
499.15
RRRXPPIDPYTNJG-UHFFFAOYSA-N
perfluorooctanesulfonamide, perfluoroctylsulfonamide, perfluorooctane sulfonamide, pfosa, heptadecafluorooctanesulphonamide, perfluorooctanesulfonic acid amide, unii-80am718fml, 1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro, deethylsulfluramie, perfluorooctanesulphonamide
69785
1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonamide
C(C(C(C(C(F)(F)S(=O)(=O)N)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F
754-91-6
C8H2F17NO2S
499.15
RRRXPPIDPYTNJG-UHFFFAOYSA-N
perfluorooctanesulfonamide, perfluoroctylsulfonamide, perfluorooctane sulfonamide, pfosa, heptadecafluorooctanesulphonamide, perfluorooctanesulfonic acid amide, unii-80am718fml, 1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro, deethylsulfluramie, perfluorooctanesulphonamide
69785
1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonamide
C(C(C(C(C(F)(F)S(=O)(=O)N)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 154.0°C to 155.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (mix of isomers) (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 499.15
Percent Purity 95%
Chemical Name or Material Perfluorooctanesulfonamide
compliance-icons compliance-icons
Product Identifier
  • Perfluorooctanesulfonamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'