missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 1.679 Millesimal Buffer Solution Sachets, Hanna Instruments™
Click to view available options
Quantity:
20 mL
Packaging:
25Pack
Description
- Premium quality pH 1.679 millesimal buffer solution that is NIST traceable and supplied with a Certificate of Analysis
- Millesimal buffer solutions are designed for customers who require a resolution of 0.001 for their pH measurements
- Temperature chart of the actual pH value at various temperatures printed on each sachet
- Produced in accordance with ISO 3696/BS3978 standards using high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.002 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
Specifications
Color | Undesignated |
Concentration | >1 to <5% |
pH | 1.67 |
Physical Form | Liquid |
Quantity | 20 mL |
Formulation | Potassium Tetraoxalate Dihydrate |
Odor | Odorless |
Packaging | 25Pack |
Traceability to NIST | Yes |
Safety and Handling
Product Identifier
- pH 1.679 Buffer Solution
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : potassium tetraoxalate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'