missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ pH Indicator Paper Strips
Dip the pH test paper into the solution to be tested and compare the colour with the colour chart provided. Fisherbrand™ pH Indicator Paper Strips provide quick and easy pH determination.
4.58 € - 5.36 €
Specifications
Format | Strip |
---|---|
Includes | Color indicator chart |
Quantity | 200/Pk. |
Type | pH Test Strip |
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
15575669
|
Fisherbrand™
|
0 to 11 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15525679
|
Fisherbrand™
|
0 to 14 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Includes color chart label for easy reference and pH level determination
- 200 strips
Specifications
Strip | |
Color indicator chart | |
200/Pk. | |
pH Test Strip |