Learn More
Fisherbrand™ pH Qualitative Indicator Paper, Without Color Chart
Provides simple acid/base determinations at a set pH transition point. Fisherbrand™ pH Qualitative Indicator Paper, Without Color Chart are simple, completely impregnated indicator papers used to indicate if the pH of a solution is above or below the transition range (color change).
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.05760kg
Description
Test papers allow the easy determination of an alkaline or acidic environment. Professional analysts as well as chemical laymen appreciate these tests. The paper rolls are supplied in plastic dispensing reels for easy use: just pull out and tear off a strip.
- Easy: Sophisticated chemical determinations are simplified to a Dip & Read process
- Rapid: Immediate results for on-site analysis
- Accurate: High quality papers ensure optimal results
- Stable: Supplied in plastic reels to protect against environmental conditions
Specifications
Red to Blue | |
Determining a strongly acidic solution | |
Transition point: 5.0 - 3.0 pH | |
Congo Paper MN 816 N | |
7 mm |
Roll | |
1 Dispenser, 4 Indicator paper rolls | |
4 Each | |
5 m |
Safety and Handling
- Kongopapier MN 816 N
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Congo red (pH indicator)