missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Red Litmus Paper Reel
Provides a general test for acidic or alkaline reactions. Fisherbrand™ Red Litmus Paper Reels are comprised of completely impregnated indicator paper.
Brand: Fisherbrand
Code : NEW
Additional Details : Weight : 0.06204kg
Description
- Color change from red to blue
- Store in a cool, dry place
- Avoid exposure to sunlight and moisture
- Serves in neutralization reactions to check whether the change from acid to alkaline or vice versa has taken place
Specifications
Red to Blue | |
Reel | |
1 Each | |
5000 mm |
5 to 8 | |
Determination of acidic and alkaline reaction | |
pH Paper | |
7 mm |