Learn More
pH Singles Calibration Solutions in 20mL Pouches, Oakton™
Catalog Number 11965484
Click to view available options
pH:
4.01
10.00
Economical pouches are accurate and convenient.
The pH Singles are ideal for field calibration—just select the pouches needed and carry them to your test site for on-the-spot calibration and verification. No bottles to carry or clean. Pouches are air-tight for maximum freshness and unquestionable reliability. Packets are labeled with temperature versus pH reference tables. Sold in packs of twenty 20mL pouches.
Certifications
NIST traceable, All solutions are certified traceable to NIST™ standards.
TRUSTED_SUSTAINABILITY
pH 10.00
Quantity 20 mL Pouches