Learn More
Motic™ Phase Centering Telescope
Use phase contrast to improve visualization of transparent specimens. Motic™ Phase Centering Telescope allows you to view a fixed phase ring for condenser alignment.
Brand:  Motic™ 12220280
Additional Details : Weight : 0.01000kg
Product Code. 15405132
  • 98.00 € / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
  • Use to align annulus and objective for proper phase contrast viewing.
  • Improve visualization of transparent specimens.
Specifications
Specifications
DMW-BI, B3, B1, B1-223ASC, B3-223PL Microscopes
23.2 mm