missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenacaine Hydrochloride, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Phenacaine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Phenacaine Hydrochloride |
| CAS | 620-99-5 |
| Analyte or Component Names | Phenacaine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ethanimidamide, N,N'-bis(4-ethoxyphenyl)-, hydrochloride (1:1), Acetamidine, N,N'-bis(p-ethoxyphenyl)-, hydrochloride (7CI), Acetamidine, N,N'-bis(p-ethoxyphenyl)-, monohydrochloride (8CI), Ethanimidamide, N,N'-bis(4-ethoxyphenyl)-, monohydrochloride (9CI), Holocaine hydrochloride, Phenacaine hydrochloride |
| InChI Formula | InChI=1S/C18H22N2O2.ClH/c1-4-21-17-10-6-15(7-11-17)19-14(3)20-16-8-12-18(13-9-16)22-5-2;/h6-13H,4-5H2,1-3H3,(H,19,20);1H |
| SMILES | Cl.CCOc1ccc(N\C(=N\c2ccc(OCC)cc2)\C)cc1 |
| IUPAC Name | N,N'-bis(4-ethoxyphenyl)ethanimidamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction