Learn More
Fisher Bioreagents Phenol / Chloroform / Isoamyl alcohol pH 8 25:24:1 ratio for extraction of proteins from crude nucle keep cool - store at 0°C to 5°C Fisher BioReagents
Brand:  Fisher Bioreagents 40-1200-10
Phenol / Chloroform / Isoamyl alcohol pH 8 25:24:1 ratio for extraction of proteins from crude nucle keep cool - store at 0°C to 5°C Fisher BioReagents
Code : 32
Additional Details : Weight : 0.10000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : II
Product Code. 12777119