Learn More
Phenol Liquified 80% (w/w) In Water, Certified AR, for Analysis, Fisher Chemical™
C6H6O, CAS Number-108-95-2, benzenol, carbolic acid, hydroxybenzene, monophenol, oxybenzene, phenic acid, phenyl hydrate, phenyl hydroxide, phenylic acid, phenylic alcohol, 2.5L, CHEBI:15882, Red, 94.0414g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 40.5 deg.C, 2143, 94.113, Plastic Coated Glass bottle
Brand: Fisher Chemical P/2316/PB17
140.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 22
Additional Details : CAS Number : 108-95-2 Weight : 2.67500kg Transport : UN number : 2821 Chem class : 6.1 Pack group : II
Description
C6H6O, CAS Number-108-95-2, benzenol, carbolic acid, hydroxybenzene, monophenol, oxybenzene, phenic acid, phenyl hydrate, phenyl hydroxide, phenylic acid, phenylic alcohol, 2.5L, CHEBI:15882, Red, 94.0414g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 40.5 deg.C, 2143, 94.113, Plastic Coated Glass bottle
Specifications
108-95-2 | |
Red | |
C6H6O | |
2.5 L | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
phenol | |
3.2 | |
CHEBI:15882 | |
1mmHg 40 |
40.5°C | |
Plastic Coated Glass bottle | |
MFCD00002143 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
OC1=CC=CC=C1 | |
94.11 | |
996 | |
94.0414g/mol | |
Liquid |
Safety and Handling
- Phenol liquified
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.