missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phenol, Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer

Brand:  Alfa Aesar™ J67050.AE

Code : NEW

Additional Details : Weight : 0.00010kg Transport : UN number : 2821 Chem class : 6.1 Pack group : II

Quantity 100mL

 View more versions of this product

Product Code. 15474495

  • 97.10 € / 100mL

Estimated Shipment date
from Supplier 24-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

108-95-2
94.113
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
996
phenol
C6H6O
MFCD00002143
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
CHEBI:15882
C1=CC=C(C=C1)O

Specifications

Phenol
108-95-2
MFCD00002143
1.060g/mL
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
996
94.11
Undesignated
Light Sensitive
Ready-to-Use, buffer saturated solution pH 6.6, with alkaline buffer
C6H6O
UN2821
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
C1=CC=C(C=C1)O
94.113
CHEBI:15882
Liquid
100mL
Videos
SDS