missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5391557
Click to view available options
Quantity:
5 g
25 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
143-74-8
C19H14O5S
354.38
MFCD00003552
BELBBZDIHDAJOR-UHFFFAOYSA-N
4766
CHEBI:31991
4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
This item is not returnable. View return policy
143-74-8
C19H14O5S
354.38
MFCD00003552
BELBBZDIHDAJOR-UHFFFAOYSA-N
4766
CHEBI:31991
4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

High quality chemicals for laboratory and specialized industrial use

Formula Weight 354.39
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 5 g
Chemical Name or Material Phenol Red
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'